Skip to Content
Merck
All Photos(1)

Documents

PHR1245

Supelco

Vanillin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
Beilstein:
472792
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. V0050000
traceable to USP 1710006

vapor density

5.3 (vs air)

vapor pressure

>0.01 mmHg ( 25 °C)

API family

vanillin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards.
Vanillin is an aromatic compound, and is used for flavoring foods. It is has biogenetic relationship to phenylpropanoid pathway. It is mostly used in foods, beverages, perfumes and pharmaceuticals.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Vanillin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3259 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanillin
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 6037-6037 (2013)
Biotechnological production of vanillin.
Priefert, H., J. Rabenhorst, and A. Steinbuchel
Applied Microbiology and Biotechnology, 56.3-4, 296-314 (2001)
Igor A O Reis et al.
Food chemistry, 135(4), 2453-2461 (2012-09-18)
A single-step selective separation of two food additives was investigated using alcohol-salt aqueous two-phase systems (ATPS). The selective partitioning of two of the most used additives from a processed food waste material, vanillin and l-ascorbic acid, was successfully accomplished. The
Yavuz Yardım et al.
Food chemistry, 141(3), 1821-1827 (2013-07-23)
A method for the determination of food additive vanillin was developed by adsorptive stripping voltammetry. Its determination was carried out at the anodically pre-treated boron-doped diamond electrode in aqueous solutions. Using square-wave stripping mode, the compound yielded a well-defined voltammetric
Ana Rita Brochado et al.
Biotechnology and bioengineering, 110(2), 656-659 (2012-09-26)
Overproduction of a desired metabolite is often achieved via manipulation of the pathway directly leading to the product or through engineering of distant nodes within the metabolic network. Empirical examples illustrating the combined effect of these local and global strategies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service