Skip to Content
Merck
All Photos(1)

Key Documents

PHL80822

Linarin

phyproof® Reference Substance

Synonym(s):

5,7-Dihydroxy 4′-methoxyflavone 7-rutinoside, 7-((6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl)oxy)-5-hydroxy-2-(4-methoxyphenyl)-4H-benzopyran-4-one, Acacetin-7-rutinoside, Buddleoside, Linarigenin glycoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32O14
CAS Number:
Molecular Weight:
592.55
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

O1[C@H]([C@@H]([C@@H]([C@H]([C@@H]1C)O)O)O)OC[C@H]2O[C@H]([C@@H]([C@H]([C@@H]2O)O)O)Oc3cc4[o]c(c[c](c4c(c3)O)=O)c5ccc(cc5)OC

InChI

1S/C28H32O14/c1-11-21(31)23(33)25(35)27(39-11)38-10-19-22(32)24(34)26(36)28(42-19)40-14-7-15(29)20-16(30)9-17(41-18(20)8-14)12-3-5-13(37-2)6-4-12/h3-9,11,19,21-29,31-36H,10H2,1-2H3/t11-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

YFVGIJBUXMQFOF-PJOVQGMDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Linarin as a Neuroprotective Agent Studies: Linarin demonstrated potential neuroprotective effects, ameliorating ischemia-reperfusion injury by inhibiting endoplasmic reticulum stress targeting AKR1B1, which suggests its applicability in the development of treatments for neurodegenerative disorders (Zhang et al., 2024).

  • Anti-inflammatory Effects of Linarin in Biochemistry: The anti-inflammatory properties of Linarin make it a significant compound for research in pharmacological sciences, as it might lead to new insights into the modulation of inflammatory pathways in chronic diseases (Cheriet et al., 2024).

  • Linarin in Chromatography and Spectrometry Analysis: As a chromatography standard, Linarin is essential for developing analytical methods that ensure the quality control of medicinal plants and herbal products, which is key for natural product research and drug development (Gontar et al., 2024).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo-Il Kim et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 79, 153351-153351 (2020-09-29)
Since enhanced bone resorption due to osteoclast differentiation and activation cause skeletal diseases, there is a growing need in therapeutics for combating bone-resorbing osteoclasts. Botanical antioxidants are being increasingly investigated for their health-promoting effects on bone. Edible Cirsium setidens contains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service