Skip to Content
Merck
All Photos(1)

Key Documents

P5239

Supelco

Pinocembrin

analytical standard, ≥95.0% (HPLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O4
CAS Number:
Molecular Weight:
256.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

194-195 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC1=C(C(C[C@@H](C2=CC=CC=C2)O3)=O)C3=CC(O)=C1

InChI

1S/C15H12O4/c16-10-6-11(17)15-12(18)8-13(19-14(15)7-10)9-4-2-1-3-5-9/h1-7,13,16-17H,8H2/t13-/m0/s1

InChI key

URFCJEUYXNAHFI-ZDUSSCGKSA-N

General description

Anti-inflammatory.
Pinocembrin is a major flavonoid molecule, isolated from a variety of plants such as Euphorbia, Sparattosperma leucanthum, Populus, Eucalyptus, Pinus heartwood, etc. It exhibits antitumor, antimicrobial, anti-inflammatory and antioxidant properties.[1][2]

Application

Pinocembrin has been used as a standard in the determination of pinocembrin in propolis extract, analyzed using ultraviolet-visible spectrophotometry (UV-Vis).[3]
Pinocembrin may be used as an analytical reference standard for the quantification of the analyte in honey samples using capillary electrophoresis technique.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Protect from light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Liu et al.
BMC medicine, 10, 105-105 (2012-09-20)
It is known that amyloid-β peptide (Aβ) plays a pivotal role in the pathogenesis of Alzheimer's disease (AD). Interaction between Aβ and the receptor for advanced glycation end products (RAGE) has been implicated in neuronal degeneration associated with this disease.
Pinocembrin: a novel natural compound with versatile pharmacological and biological activities
Rasul A, et al.
BioMed Research International, 2013(1-2), 71-78 (2013)
María de Jesús González-Búrquez et al.
Evidence-based complementary and alternative medicine : eCAM, 2018, 7092416-7092416 (2018-09-04)
Propolis is a resin that honey bees (Apis mellifera) produce by mixing wax, exudates collected from tree shoots, pollen, and enzymes. It has been used for its biological properties against pathogenic microorganisms including those of viral origin. In the present
Capillary electrophoresis as an alternative to HPLC for determination of honey flavonoids
Delgado.C, et al.
Chromatographia, 38(1-2), 71-78 (1994)
Xu Yang et al.
Biomolecules, 8(2) (2018-06-21)
Pinocembrin is a natural flavonoid compound which is capable of antioxidant, antibacterial, anti-inflammatory, and antineoplastic activities. The present study aimed to enhance the solubility and antioxidant activities of pinocembrin by complex formation with lecithin. The physicochemical characteristics of pinocembrin⁻lecithin complex

Articles

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service