Skip to Content
Merck
All Photos(1)

Key Documents

P3015

Sigma-Aldrich

Poly(ethylene glycol)

average mol wt 200

Synonym(s):

Polyethylene glycol, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

form

liquid

Quality Level

mol wt

average mol wt 200

pH

4.5-7.5

viscosity

4.3 cSt(210 °F)(lit.)

mp

-65 °C (lit.)

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly (ethylene glycol) (PEG) is non-ionic, hydrophilic and non-toxic polymer. It is widely used in the emerging field of polymer‐based drug delivery.[1] PEG is available in different molecular weights. It facilitates the purification and crystal growth of proteins and nucleic acids. PEG and dextran together result in aqueous polymer two phase system, which is required for the purification of biological materials. It also interacts with cell membrane, thereby allowing cell fusion.[2]

Application

Poly(ethylene glycol) has been used in the preparation of methyl cellulose (MC)-based polymer electrolytes.[3] It has also been used as a solvent to analyse the molecular weight cut off and rejection studies of nanofiltration membranes.[4]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristics of nanofiltration membranes in aqueous ethanol
Tsui EM and Cheryan M
Journal of Membrane Science , 237(1-2), 61-69 (2004)
Electrical double-layer capacitors with plasticized polymer electrolyte based on methyl cellulose
Shuhaimi NEA, et al.
Polymer Bull., 69(7), 807-826 (2012)
Poly(Ethylene Glycol) Chemistry: Biotechnical and Biomedical Applications (2013)
Yi-Zhong Gu et al.
Toxicological sciences : an official journal of the Society of Toxicology (2019-08-14)
MK-7680, a cyclic nucleotide prodrug, caused significant kidney tubule injury in female rats when administered orally at 1000 mg/kg/day for 2 weeks using 10% Polysorbate 80 as vehicle. However, kidney injury was absent when MK-7680 was administered at the same dose
Poly (ethylene glycol) in drug delivery: pros and cons as well as potential alternatives.
Knop K, et al.
Angewandte Chemie (International Edition in English), 49(36), 6288-6308 (2010)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. What is the concentration of this product?

    1 answer
    1. The product P3015 is a neat liquid with a density of 1.1239 g/ml at 20°C. Assuming it is pure and has a molecular weight of 200, the molarity of this product is 5.62 M.

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    0 answers

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service