Skip to Content
Merck
All Photos(1)

Key Documents

NIST679

Brick clay

NIST® SRM® 679

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 75 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in the determination of constituent elements in clay or materials of a similar matrix. SRM 679 consists of brick clay that ws air dried, ball milled and passed through a 200 mesh sieve. Gor more information, please refer to the SDS and COA.

SRM 679_cert

SRM 679_SDS

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Calcium (Ca), Iron (Fe), Magnesium (Mg), Potassium (K), Silicon (Si), Sodium (Na), Titanium (Ti), Barium (Ba), Chromium (Cr), Lithium (Li), Strontium (Sr)

Reference/Informational value is provided for the following analytes.
Cerium (Ce), Cesium (Cs), Cobalt (Co), Europium (Eu), Hafnium (Hf), Manganese (Mn), Phosphorus (P), Rubidium (Rb), Scandium (Sc), Thorium (Th), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service