Skip to Content
Merck
All Photos(1)

Key Documents

NIST1766

Low alloy steel

NIST® SRM® 1766

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in the form of a disk, 34 mm (1 3/8 in.) and 19 mm (3/4 in.) thick. It is intended for use in optical emission and x-ray spectrometric methods of analysis. For more information, plese refer to the SDS and COA.

SRM 1766_cert

SRM 1766_SDS

Application

The Standard Reference Material (SRM) provides certified values that are primarily intended for validating chemical and instrumental methods used to analyze the elemental content of high-purity iron, low alloy steel, and similar ferrous alloys. These values can also be used to validate the value assignments of in-house reference materials in laboratories.

Features and Benefits

  • Certified Values for various elements in SRM 1766 are provided and the values are metrologically traceable to SI unit of mass fraction, expressed as percentage.
  • Non-certified value for boron is provided.
  • Maintenance of certified and non-certified values, period of validity, instructions for storage, and use are provided on NIST certificate

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Nickel (Ni), Antimony (Sb), Niobium (Nb), Arsenic (As), Nitrogen (N), Carbon (C), Phosphorus (P), Chromium (Cr), Silicon (Si), Cobalt (Co), Silver (Ag), Copper (Cu), Sulfur (S), Lead (Pb), Tin (Sn), Manganese (Mn), Titanium (Ti), Molybdenum (Mo), Vanadium (V)

Reference/Informational value is provided for the following analytes.
Boron (B), Bismuth (Bi), Tantalum (Ta), Cerium (Ce), Tellurium (Te), Magnesium (Mg), Tungsten (W), Selenium (Se), Zirconium (Zr)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service