Skip to Content
Merck
All Photos(1)

Key Documents

FPPMCC1

Flash Point Standard - Pensky Martens

certified reference material, ASTM D 93 Proc. A (59.0 °C)

Synonym(s):

Pensky-Martens flash point calibration standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

according to ASTM® D93

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Flash Point Standards are tested and certified in accordance with ASTM D93 ′Standard Test Method for Flash Point by Pensky Martens Closed Cup Tester - Procedure A′. Procedure A is applicable to distillate fuels - diesel, biodiesel blends, kerosene, heating oil, turbine fuels and lubricating oils. These UKAS certified standards are specifically designed to be used for regular instrument performance checks.
Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Pensky Martens Flash Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Application

Flash point standard-pensky martens closed cup tester may be used as a standard, to determine the lower flash point for two binary systems comprising of water/acetic acid and water/propionic acid.

Features and Benefits

  • Secondary working standard is categorized in accordance with ASTM D93 Procedure A for equipment performance checks.
  • Packaged in 3 X 80mL bottles - ′Single shot′; one bottle per test ensures zero contamination / high end volatile loss.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper-evident security packaging.
  • Shelf life of 12 months.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The lower flash points of binary systems containing non-flammable component
Lee S and Ha M-D.
Korean Journal of Chemical Engineering, 20(5), 799-802 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service