Skip to Content
Merck
All Photos(1)

Key Documents

CVCO15W40

Certified Viscosity Check Oil

15W40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

General description

Manufactured and certified in strict accordance with the requirements of ISO 17025 and ISO 17034. Kinematic viscosity measurements have been made in accordance with ASTM D445, using Reference Viscometers certified in accordance with ASTM D446 and ASTM D2162. Density, Cold-Crank Simulator (CCS) and Viscosity Index measurements have been made in accordance with ASTM D4052, ASTM D5293 and ASTM D2270 respectively.

The viscosity values for the certified viscosity check oils have been tested and certified at typical blend targets. These check oils are for the validating the performance of Cold-Cranking Simulator and Kinematic Viscometers by measuring commercially available formulated engine oils.

Please note you can also see information about this product on the Paragon Scientific webpage

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service