Skip to Content
Merck
All Photos(1)

Key Documents

CRMFAGO

Certified Reference Material Fatty Acid Methyl Ester (FAME) Standard

Diesel

Synonym(s):

Diesel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

format

matrix material

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Fatty Acid Methyl Ester (FAME) Certified Reference Material for the calibration and verification of laboratory instruments used for the determination of Fatty Acid Methyl Ester (FAME) content in middle distillates - Infrared spectrometry method, EN 14078

Application


  • FAME reference standard for lipid profiling: The Certified Reference Material Fatty Acid Methyl Ester (FAME) Standard is extensively used in lipid profiling to ensure accuracy and reliability in the quantification of fatty acids in biological samples, supporting advancements in pharmaceutical research and nutritional studies (Gallego et al., 2018).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

134.6 °F

Flash Point(C)

57 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service