Skip to Content
Merck

CRMAPKR

Aniline Point Standard

ASTM D 611 (62.30°C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41116107
EC Number:
294-799-5
NACRES:
NA.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Aniline Point Standard, ASTM D 611 (62.30°C)

grade

certified reference material

agency

according to ASTM® 611

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

bp

90-300 °C

mp

-49 °C

application(s)

petroleum

format

mixture

Quality Level

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
CRMCNGOCRMUCPGOCRMUDIKR
format

mixture

format

mixture

format

mixture

format

-

bp

90-300 °C

bp

141-462 °C

bp

141-462 °C

bp

90-300 °C

mp

-49 °C

mp

-

mp

-

mp

-49 °C

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

application(s)

petroleum

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Fuels Testing Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

84.2 - 158.0 °F - closed cup

flash_point_c

29 - 70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service