Skip to Content
Merck
All Photos(3)

Key Documents

C5080

Sigma-Aldrich

Calcium chloride dihydrate

BioXtra, ≥99.0%

Synonym(s):

CaCl2.2H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 M, clear, colorless
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

176 °C (dec.) (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ba: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.005%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Optimization of guar gum-based anti-browning coating for prolonging the shelf life of cut potatoes.: Calcium chloride dihydrate is utilized in developing an anti-browning coating for cut potatoes, significantly extending their shelf life. This application showcases its effectiveness in food preservation, ensuring quality and reducing food waste (Mondal et al., 2024).

  • Phosphatidylinositol-specific phospholipase C-associated phospholipid metabolism mediates DcGLRs channel to promote calcium influx under CaCl(2) treatment in shredded carrots during storage.: This study explores how calcium chloride dihydrate influences phospholipid metabolism and calcium influx in shredded carrots, enhancing their quality during storage. The research provides insights into the biochemical pathways involved in maintaining post-harvest vegetable quality (Zhang et al., 2024).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaina R Eckhouse et al.
Circulation, 128(11 Suppl 1), S186-S193 (2013-10-18)
Thoracic aortic aneurysms (TAAs) develop secondary to abnormal aortic extracellular matrix remodeling, resulting in a weakened and dilated aortic wall that progressed to rupture if left unattended. Currently, no diagnostic/prognostic tests are available for the detection of TAA disease. This
Enrique Jambrina et al.
Journal of biomolecular screening, 21(5), 468-479 (2016-02-04)
N-methyl-D-aspartate receptors (NMDARs) are ionotropic glutamate receptors that play an important role in synaptic plasticity and learning and memory formation. Malfunctioning of NMDARs, in particular the reduction in NMDAR activity, is thought to be implicated in major neurological disorders. NMDAR
Melina J Scholze et al.
Development (Cambridge, England), 145(11) (2018-05-05)
Asymmetric division is crucial for embryonic development and stem cell lineages. In the one-cell Caenorhabditis elegans embryo, a contractile cortical actomyosin network contributes to asymmetric division by segregating partitioning-defective (PAR) proteins to discrete cortical domains. In the current study, we
Lilith M Caballero Aguilar et al.
Journal of colloid and interface science, 587, 240-251 (2020-12-29)
Drug delivery systems such as microspheres have shown potential in releasing biologicals effectively for tissue engineering applications. Microfluidic systems are especially attractive for generating microspheres as they produce microspheres of controlled-size and in low volumes, using micro-emulsion processes. However, the
Minho Noh et al.
Cell systems, 5(4), 418-426 (2017-10-02)
Escherichia coli gene expression knockdown using synthetic small RNA (sRNA) can be fine-tuned by altering sRNA sequences to modulate target mRNA-binding ability, but this requires thorough checking for off-target effects. Here, we present an sRNA gene expression knockdown system fine-tuned

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service