Skip to Content
Merck
All Photos(1)

Key Documents

BCR298

2,2′,3,4,4′,5,5′-Heptachlorobiphenyl (IUPAC No. 180)

BCR®, certified reference material

Synonym(s):

2,2′,3,4,4′,5,5′-Heptachlorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H3Cl7
CAS Number:
Molecular Weight:
395.32
Beilstein:
2509255
Ballschmiter Number:
180
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1cc(Cl)c(cc1Cl)-c2cc(Cl)c(Cl)c(Cl)c2Cl

InChI

1S/C12H3Cl7/c13-6-3-8(15)7(14)1-4(6)5-2-9(16)11(18)12(19)10(5)17/h1-3H

InChI key

WBHQEUPUMONIKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR298

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elsa C Antunes-Fernandes et al.
Toxicology letters, 206(2), 158-165 (2011-07-26)
Traditional risk assessment of potential endocrine-disruptive pollutants, including PCBs, focus mainly on the effects of parent compounds. Still, biotransformation results in systemic exposure to PCBs and their bioactive metabolites. In the present paper, the effects of twenty ultra-pure non-dioxin-like (NDL)
Joanne S Colt et al.
Blood, 113(9), 1899-1905 (2008-12-11)
Organochlorine exposure was linked to non-Hodgkin lymphoma (NHL) risk. To determine whether this relation is modified by immune gene variation, we genotyped 61 polymorphisms in 36 immune genes in 1172 NHL cases and 982 controls from the National Cancer Institute-Surveillance
Robert Roos et al.
Toxicology, 284(1-3), 42-53 (2011-04-05)
PCB 180 (2,2',3,4,4',5,5'-heptachlorobiphenyl) is a persistent and accumulating polychlorinated biphenyl abundantly present in food and the environment. In this study, we used highly purified PCB 180 (dioxinlike impurities: 2.7 ng TEQ(WHO)/g PCB 180) in a 28-day toxicity study in young
Roshan Tofighi et al.
Toxicological sciences : an official journal of the Society of Toxicology, 124(1), 192-201 (2011-09-13)
Developmental exposure to food contaminants, such as polychlorinated biphenyls (PCBs), has been considered as a possible cause of neurodevelopmental disorders. We have investigated the effects of noncytotoxic concentrations of PCBs 153 and 180 on spontaneous differentiation of rat embryonic neural
Johan Maervoet et al.
Environmental toxicology and chemistry, 24(3), 597-602 (2005-03-23)
Fertilized chicken eggs were injected with high doses of individual polychlorinated biphenyl (PCB) congeners (0.5 microg of PCB 77, 9.8 microg of PCB 153, or 10.9 microg of PCB 180) before incubation to investigate the structure-specific uptake of these compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service