Skip to Content
Merck
All Photos(2)

Key Documents

93791

Supelco

Allantoin

analytical standard

Synonym(s):

5-Ureidohydantoin, Glyoxylic(acid) diureide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3
CAS Number:
Molecular Weight:
158.12
Beilstein:
102364
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

ign. residue

≤0.2%

mp

230 °C (dec.) (lit.)

solubility

H2O: 0.1 g/10 mL, clear, colorless

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)NC1NC(=O)NC1=O

InChI

1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)

InChI key

POJWUDADGALRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allantoin may be used as an analytical reference standard for the quantification of the analyte in biological samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Purine metabolite via the uric acid pathway. Uric acid also reacts with free radicals to produce allantoin, thus allantoin may be a useful biomarker for oxidative stress.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of oxonic acid, uric acid, creatine, allantoin, xanthine and hypoxanthine in poultry litter by reverse phase HPLC
Eiteman.A.M, et al.
Fresenius Journal of Analytical Chemistry, 348(10), 680-683 (1994)
Improved HPLC method for the simultaneous determination of allantoin, uric acid and creatinine in cattle urine
George.K.S, et al.
Journal of Chromatography. B, Biomedical Applications, 832(1), 134-137 (2006)
Measurement of allantoin in urine and plasma by high-performance liquid chromatography with pre-column derivatization
Chen.B.X, et al.
Journal of Chromatography. B, Biomedical Applications, 617(2), 241-247 (1993)
Camila G Moreira et al.
Journal of ethnopharmacology, 141(3), 1005-1011 (2012-04-17)
Pyrostegia venusta is a native Brazilian plant which has a variety of uses in traditional folk medicine including the treatment of vitiligo. However, its effectiveness on melanogenesis is not yet elucidated. This study aimed to investigate the melanogenic activity of
Peng Wang et al.
Journal of agricultural and food chemistry, 60(11), 2793-2798 (2012-03-01)
Despite increasing knowledge of allantoin as a phytochemical involved in rice, relatively little is known about its distribution and function in rice grains. In this study, allantoin was quantified in 15 Chinese rice grains, and its contents varied with grain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service