Skip to Content
Merck
All Photos(1)

Key Documents

918709

Sigma-Aldrich

5,6-Dihydroxyindole

Synonym(s):

Dopamine lutine, Indolylquinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NO2
CAS Number:
Molecular Weight:
149.15
MDL number:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

Quality Level

storage temp.

2-8°C

SMILES string

OC1=CC(C=CN2)=C2C=C1O

InChI

1S/C8H7NO2/c10-7-3-5-1-2-9-6(5)4-8(7)11/h1-4,9-11H

InChI key

SGNZYJXNUURYCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service