Skip to Content
Merck
All Photos(1)

Key Documents

90941

Supelco

[(3R)-3-Hydroxybutyryl]-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-2-[(3R)-3-hydroxy-1-oxobutoxy]-N,N,N-trimethyl-1-propanaminium inner salt, [(R)-3-Hydroxybutyryl]-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NO5
CAS Number:
Molecular Weight:
247.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

solid

optical activity

[α]/D -40.0 to -34.0°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

impurities

≤15% water (calculated from elemental analysis)

storage temp.

−20°C

SMILES string

C[N+](C)(C)C[C@H](OC(C[C@H](O)C)=O)CC([O-])=O

InChI

1S/C11H21NO5/c1-8(13)5-11(16)17-9(6-10(14)15)7-12(2,3)4/h8-9,13H,5-7H2,1-4H3/t8-,9-/m1/s1

InChI key

UEFRDQSMQXDWTO-RKDXNWHRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service