Skip to Content
Merck
All Photos(3)

Key Documents

909165

Sigma-Aldrich

Hoveyda-Grubbs Catalyst® M711

Umicore

Synonym(s):

Hoveyda-Grubbs Catalyst® M71 SIPr, [1,3-Bis(2,6-diisopropylphenyl)-2-imidazolidinylidene]dichloro[(2- isopropoxy)(5-trifluoroacetamido)benzylidene]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H50N3Cl2F3O2Ru
CAS Number:
Molecular Weight:
821.80
UNSPSC Code:
12352101
NACRES:
NA.22

Application

Umicore Hoveyda-Grubbs Catalyst M711 is a homogeneous catalyst useful for alkene metathesis, cross metathesis, ring closing metathesis, and self metathesis.

Learn more about our metathesis catalysts

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service