Skip to Content
Merck
All Photos(2)

Key Documents

90281

Supelco

Andrographolide

analytical standard

Synonym(s):

(3E,4S)-3-[2-[(1R,4aS,5R,6R,8aS)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethylidene]dihydro-4-hydroxy-2(3H)-furanone, ent-(3β,12E,14R)-3,14,19-Trihydroxy-8(17),12-labdadien-16,15-olide, Andrographis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O5
CAS Number:
Molecular Weight:
350.45
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H](C\C=C3/[C@H](O)COC3=O)C(=C)CC[C@H]12

InChI

1S/C20H30O5/c1-12-4-7-16-19(2,9-8-17(23)20(16,3)11-21)14(12)6-5-13-15(22)10-25-18(13)24/h5,14-17,21-23H,1,4,6-11H2,2-3H3/b13-5+/t14-,15-,16+,17-,19+,20+/m1/s1

InChI key

BOJKULTULYSRAS-OTESTREVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Andrographolide is a major bioactive component of Andrographis paniculata Nees. which is a popular Chinese and Indian medicine.[1] It belongs to the terpenoid class of compounds.[2]

Application

Andrographolide may be used as a reference standard for the determination of andrographolide in:
  • Leaves of Andrographis paniculata by soxhlet extraction-recrystallization-column chromatography purification followed by analyses using reversed phase high performance liquid chromatography (RP-HPLC) as well as gas chromatography-mass spectrometry (GC-MS).[2]
  • Roots of Andrographis paniculata by HPLC with diode array detection (DAD), gas chromatography-mass spectrometry (GC-MS) and LC coupled to tandem mass spectrometry (MS/MS) equipped with electrospray ionization (ESI) and multiple reaction monitoring (MRM) mode of detection.[3]
  • Stems of Andrographis paniculata by ultra-HPLC combined coupled to triple quadrupole-linear ion trap mass spectrometry (QqQLIT-ESI-MS/MS) with MRM detection.[4]
  • Rat plasma samples by protein precipitation and subsequent ESI-LC-MS/MS determination using MRM detection.[5]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of phytochemicals, antioxidant activity and total phenolic content in Andrographis paniculata using chromatographic methods.
Kurzawa M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 995(2), 101-106 (2015)
Anti-dengue efficacy of bioactive andrographolide from Andrographis paniculata (Lamiales: Acanthaceae) against the primary dengue vector Aedes aegypti (Diptera: Culicidae).
Edwin ES, et al.
Acta Tropica, 163(2), 167-178 (2016)
Determination of six components of Andrographis paniculata extract and one major metabolite of andrographolide in rat plasma by liquid chromatography-tandem mass spectrometry
Wang J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 951(2), 78-88 (2014)
In vitro metabolism of sodium 9-dehydro-17-hydro-andrographolide-19-yl sulfate in rat liver s9 by liquid chromatography-mass spectrometry method.
Zheng D, et al.
Pharmacognosy magazine, 12(2), S102-S102 (2016)
Rapid quantitative analysis of multi-components in Andrographis paniculata using UPLC-QqQLIT-MS/MS: Application to soil sodicity and organic farming.
Chandra P, et al.
Industrial Crops and Products, 83(2), 423-430 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service