Skip to Content
Merck
All Photos(1)

Key Documents

87783

Supelco

Redox standard

650 mV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

description

redox potential: 650 mV ±5 mV, 25°C

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
pharmaceutical

format

mixture

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This Redox standard has a potential of 650 mV ±5 mV. It refers to the potential of the platinum electrode versus the Ag/AgCl reference electrode in 3M potassium chloride.

Application

Redox standard acts as a control standard rather than a calibration standard which controls the functionality of the sensing and reference electrodes.

Features and Benefits

  • Broad range of values
  • Very high specifications (±5mV)
  • Detailed safety data sheet (SDS) available
  • Certified with proven verifiable accuracy and uncertainty of measurement
  • Guaranteed stability throughout its entire shelf life

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service