Skip to Content
Merck
All Photos(1)

Key Documents

86470

Sigma-Aldrich

p-Terphenyl

suitable for scintillation, ≥98.5% (HPLC)

Synonym(s):

1,4-Diphenylbenzene, PT, PTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4C6H5
CAS Number:
Molecular Weight:
230.30
Beilstein:
1908447
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.95 (vs air)

Quality Level

Assay

≥98.5% (HPLC)

form

solid

bp

389 °C (lit.)

mp

212-213 °C (lit.)
212-214 °C

fluorescence

λex 286 nm; λem 343 nm in DMF

λ

in ethanol

UV absorption

λ: 277 nm Amax: ≥98.5%

suitability

suitable for scintillation

SMILES string

c1ccc(cc1)-c2ccc(cc2)-c3ccccc3

InChI

1S/C18H14/c1-3-7-15(8-4-1)17-11-13-18(14-12-17)16-9-5-2-6-10-16/h1-14H

InChI key

XJKSTNDFUHDPQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

λmax. 277 nm, log ε 4.50

Analysis Note

In addition to the emission maximum at 343 nm, there are lower maxima at 325 and 327 nm.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurélia Caputo et al.
Biology direct, 10, 5-5 (2015-04-19)
Alterations in gut microbiota composition under antibiotic pressure have been widely studied, revealing a restricted diversity of gut flora, including colonization by organisms such as Enterococci, while their impact on bacterial load is variable. High-level colonization by Akkermansia muciniphila, ranging
Thomas A Lanz et al.
PloS one, 10(3), e0121744-e0121744 (2015-03-19)
Increased protein levels of striatal-enriched tyrosine phosphatase (STEP) have recently been reported in postmortem schizophrenic cortex. The present study sought to replicate this finding in a separate cohort of postmortem samples and to extend observations to striatum, including subjects with
Jonathan Breeze et al.
Nature communications, 6, 6215-6215 (2015-02-24)
Recently, the world's first room-temperature maser was demonstrated. The maser consisted of a sapphire ring housing a crystal of pentacene-doped p-terphenyl, pumped by a pulsed rhodamine-dye laser. Stimulated emission of microwaves was aided by the high quality factor and small
Xiaojie Zhou et al.
PloS one, 9(9), e106932-e106932 (2014-09-30)
The German cockroach, Blattella germanica, is an important insect pest that transmits various pathogens mechanically and causes severe allergic diseases. This insect has long served as a model system for studies of insect biology, physiology and ecology. However, the lack
Franziska Mußbach et al.
Journal of cancer research and clinical oncology, 141(5), 813-825 (2014-11-07)
There is growing evidence for a role of proteinase-activated receptors (PARs), a subfamily of G protein-coupled receptors, in cancer. We have previously shown that PAR1 and PAR4 are able to promote the migration of hepatocellular carcinoma (HCC) cells suggesting a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service