Skip to Content
Merck
All Photos(2)

Key Documents

85429

Supelco

Sinapic acid

Sinapic acid
1 of 1 reviewers received a sample product or took part in a promotion

matrix substance for MALDI-MS, ≥99.0% (T)

Synonym(s):

3,5-Dimethoxy-4-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxy-cinnamic acid, Sinapinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O5
CAS Number:
Molecular Weight:
224.21
Beilstein:
2699118
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.0% (T)

form

powder

analyte chemical class(es)

dendrimers, fullerenes, peptides, proteins

technique(s)

MALDI-MS: suitable

color

slightly yellow

mp

~202 °C

solubility

dioxane: 1 g/10 mL at Hot, clear to slightly hazy (Very Faint Yellow to Dark Yellow and Very Faint Orange to Dark Orange)

cation traces

Ba: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for UV test

SMILES string

COc1cc(\C=C\C(O)=O)cc(OC)c1O

InChI

1S/C11H12O5/c1-15-8-5-7(3-4-10(12)13)6-9(16-2)11(8)14/h3-6,14H,1-2H3,(H,12,13)/b4-3+

InChI key

PCMORTLOPMLEFB-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sinapic acid is a type of phenolic acid.[1] Removal of amino group from phenylalanine, catalysing with enzyme phenylalanine ammonia-lyase forms cinnamic acid. The precursor of hydroxycinnamates is produced after hydroxylation of benzene ring. One of the precursors produced is sinapic acid.[2]

Application

Sinapic acid was suitable as phenolic standard for HPLC analysis in determination of Sinapic acid derivatives in Canola extracts.[1] 10 g/L of sinapinic acid with solvent was suitable as matrix for ultraviolet laser desorption mass spectrometric determination of proteins.[3] It was also suitable to use as matrix for MALDI-TOFMS and MALDI-ion mobility-TOFMS to determine phospholipids in tissue.[4]

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.N. Singh
Climate Change and Crops, 249-249 (2009)
Shelley N Jackson et al.
Journal of the American Society for Mass Spectrometry, 16(2), 133-138 (2005-02-08)
After water, lipids are the most common biomolecules found in the brain (12%). A brief perusal of the physiology, anatomy, and pathophysiology of the brain illustrates the importance of lipids. Recent advances in mass spectrometry have allowed the direct probing
R C Beavis et al.
Rapid communications in mass spectrometry : RCM, 3(12), 432-435 (1989-12-01)
The paper reports the discovery of three new matrices for the matrix-assisted laser desorption of proteins. These new matrices (sinapinic, ferulic and caffeic acids) are cinnamic acid derivatives that have several practical advantages over the nicotinic acid matrices previously used.
Rabie Khattab et al.
Journal of the American Oil Chemists' Society, 87(2), 147-155 (2010-02-17)
A high-performance liquid chromatographic (HPLC) method with diode array detection (DAD) was used to determine the total phenolics, including sinapic acid derivatives in canola. Ten Western Canadian canola seeds, six other commodity canola seeds, their corresponding press cakes and meals
Urszula Gawlik-Dziki et al.
Journal of agricultural and food chemistry, 60(18), 4603-4612 (2012-04-17)
Phenolic acids profile and antioxidant activity of six diverse varieties of spelt are reported. Antioxidant activity was assessed using eight methods based on different mechanism of action. Phenolic acids composition of spelt differed significantly between varieties and ranged from 506.6

Related Content

Discover our wide variety of products for intact mass analysis of monoclonal antibodies, including size-exclusion columns (SEC), ion exchange columns, reverse-phase columns, HPLC buffers, MALDI matrices and standards, high-purity solvents, reagents, tools for protein sample preparation, and certified reference materials.

Discover our wide variety of products for intact mass analysis of monoclonal antibodies, including size-exclusion columns (SEC), ion exchange columns, reverse-phase columns, HPLC buffers, MALDI matrices and standards, high-purity solvents, reagents, tools for protein sample preparation, and certified reference materials.

Discover our wide variety of products for intact mass analysis of monoclonal antibodies, including size-exclusion columns (SEC), ion exchange columns, reverse-phase columns, HPLC buffers, MALDI matrices and standards, high-purity solvents, reagents, tools for protein sample preparation, and certified reference materials.

Discover our wide variety of products for intact mass analysis of monoclonal antibodies, including size-exclusion columns (SEC), ion exchange columns, reverse-phase columns, HPLC buffers, MALDI matrices and standards, high-purity solvents, reagents, tools for protein sample preparation, and certified reference materials.

See All

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Akron, Ohio
    • Reviews 2
    • Votes 0
    5 out of 5 stars.

    Excellent

    Arrived quickly, product was exactly as ordered and expected

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service