Skip to Content
Merck
All Photos(1)

Key Documents

82100

Sigma-Aldrich

Propylamine

purum, ≥99.0% (GC)

Synonym(s):

1-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
1098243
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030103
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
≥99.0% (GC)

vapor density

2 (vs air)

Quality Level

vapor pressure

4.79 psi ( 20 °C)

grade

purum

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

604 °F

expl. lim.

10.4 %

refractive index

n20/D 1.388 (lit.)
n20/D 1.388

bp

48 °C (lit.)

mp

−83 °C (lit.)

density

0.719 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCN

InChI

1S/C3H9N/c1-2-3-4/h2-4H2,1H3

InChI key

WGYKZJWCGVVSQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-31.0 °F

Flash Point(C)

< -35 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan R Gordon et al.
PloS one, 9(9), e104271-e104271 (2014-09-10)
To identify molecular alterations in prostate cancers associating with relapse following neoadjuvant chemotherapy and radical prostatectomy patients with high-risk localized prostate cancer were enrolled into a phase I-II clinical trial of neoadjuvant chemotherapy with docetaxel and mitoxantrone followed by prostatectomy.
Stephanie Constantin et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(1) (2021-01-15)
Fertility relies upon pulsatile release of gonadotropin-releasing hormone (GnRH) that drives pulsatile luteinizing hormone secretion. Kisspeptin (KP) neurons in the arcuate nucleus are at the center of the GnRH pulse generation and the steroid feedback control of GnRH secretion. However
Louis-Philippe Bernier et al.
Cell reports, 27(10), 2895-2908 (2019-06-06)
Microglia, the brain's immune cells, maintain homeostasis and sense pathological changes by continuously surveying the parenchyma with highly motile large processes. Here, we demonstrate that microglia also use thin actin-dependent filopodia that allow fast nanoscale sensing within discrete regions. Filopodia
Claire Bagnéris et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(23), 8428-8433 (2014-05-23)
Voltage-gated sodium channels are important targets for the development of pharmaceutical drugs, because mutations in different human sodium channel isoforms have causal relationships with a range of neurological and cardiovascular diseases. In this study, functional electrophysiological studies show that the
Sebastian Finger et al.
Biological chemistry, 395(7-8), 769-778 (2014-07-09)
The binding of cationic polyamines to negatively charged lipid membranes is driven by electrostatic interactions and additional hydrophobic contributions. We investigated the effect of polyamines with different number of charges and charge separation on the phase transition behavior of vesicles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service