Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
81831
Propargyl bromide solution
purum, ~80% in toluene
Synonym(s):
3-Bromo-1-propyne
About This Item
Recommended Products
grade
purum
Quality Level
form
liquid
contains
~0.3% magnesium oxide light as stabilizer
concentration
~80% in toluene
density
1.39 g/mL at 20 °C
functional group
alkyl halide
bromo
storage temp.
2-8°C
SMILES string
BrCC#C
InChI
1S/C3H3Br/c1-2-3-4/h1H,3H2
InChI key
YORCIIVHUBAYBQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- new arabinogalactan propargyl ethers with degree of substitution (DS) up to 2.8, by propargylation of arabino-3,6-galactan[3]
- conjugated polyelectrolyte with polyacetylene as the backbone and pyridinium as side groups, poly(propargyl pyridinium bromide)[4]
- benzodiazepine derivative, 4-phenyl-1-(prop-2-yn-1-yl)-1H-1,5-benzodiazepin-2(3H)-one[5]
- N(3)-propargylated 2′-deoxyuridine, which can be encorporated in the DNA[6]
- chiral oxygenated acyclic natural products, via distereoselective propargylation of α-hydroxy aldehydes[1]
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3
Target Organs
Central nervous system, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
64.4 °F - closed cup
Flash Point(C)
18 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service