Skip to Content
Merck
All Photos(1)

Key Documents

80822

Supelco

(S)-Naproxen methyl ester

pharmaceutical impurity standard

Synonym(s):

Naproxen Impurity E (PhEur), (2S)-2-(6-Methoxy-2-naphthyl)propionic acid methyl ester, Methyl (αS)-6-methoxy-α-methyl-2-naphthaleneacetate, Methyl (2S)-2-(6-methoxynaphthalen-2-yl)propanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16O3
CAS Number:
Molecular Weight:
244.29
Beilstein:
4352971
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

API family

naproxen

Assay

≥98.0% (HPLC)

optical activity

[α]/D 72.0 to 84.0°, c = 0.1 in chloroform

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

87 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H](C(OC)=O)C1=CC2=CC=C(OC)C=C2C=C1

InChI

1S/C15H16O3/c1-10(15(16)18-3)11-4-5-13-9-14(17-2)7-6-12(13)8-11/h4-10H,1-3H3/t10-/m0/s1

InChI key

ZFYFBPCRUQZGJE-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-Naproxen methyl ester is an ester of the non-steroidal, anti-inflammatory drug, (S)-naproxen[1][2]

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Enantioselective hydrolysis of racemic naproxen methyl ester by two-step acetone-treated Candida rugosa lipase.
Lee GE, et al.
Process Biochemistry (Oxford, United Kingdom), 37(3), 293-298 (2001)
Bulletin of the Chemical Society of Japan, 77(1-4) (2004)
Sammy Ferri-Borgogno et al.
Cancers, 12(9) (2020-09-25)
Background & Aims: ARID1A is postulated to be a tumor suppressor gene owing to loss-of-function mutations in human pancreatic ductal adenocarcinomas (PDAC). However, its role in pancreatic pathogenesis is not clear despite recent studies using genetically engineered mouse (GEM) models.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service