Skip to Content
Merck
All Photos(1)

Documents

79590

Sigma-Aldrich

Phosphorus pentachloride

purum p.a., ≥98.0% (AT)

Synonym(s):

Phosphorus(V) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PCl5
CAS Number:
Molecular Weight:
208.24
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

0.016 hPa ( 20 °C)

Quality Level

grade

purum p.a.

Assay

≥98.0% (AT)

form

powder or crystals

impurities

≤0.005% total of sulfur (S)

pH

1 (5 g/L)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

ClP(Cl)(Cl)(Cl)Cl

InChI

1S/Cl5P/c1-6(2,3,4)5

InChI key

UHZYTMXLRWXGPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus pentachloride is a light yellow colored phosphorous halide. It participates in the synthesis of thiazoline analogs.

Application

Phosphorus pentachloride may be used in the synthesis of following compounds:
  • benzenesulfonyl chloride
  • malonic dinitrile
  • aromatic sulfonyl chlorides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infrared and Raman Spectra of Phosphorus Pentachloride.
Wilmshurst JK and Bernstein HJ.
J. Chem. Phys. , 27(3), 661-664 (1957)
Alec Lutzke et al.
ACS applied materials & interfaces, 9(3), 2104-2113 (2017-01-10)
Nitric oxide (NO) occurs naturally in mammalian biochemistry as a critical signaling molecule and exhibits antithrombotic, antibacterial, and wound-healing properties. NO-forming biodegradable polymers have been utilized in the development of antithrombotic or antibacterial materials for biointerfacial applications, including tissue engineering
Travis Marshall-Roth et al.
Nature communications, 11(1), 5283-5283 (2020-10-21)
Iron- and nitrogen-doped carbon (Fe-N-C) materials are leading candidates to replace platinum catalysts for the oxygen reduction reaction (ORR) in fuel cells; however, their active site structures remain poorly understood. A leading postulate is that the iron-containing active sites exist
Eagleson M.
Concise Encyclopedia Chemistry, 617-617 (1994)
Synthetic studies of thiazoline and thiazolidine-containing natural products-1. Phosphorus pentachloride-mediated thiazoline construction reaction.
Ino A and Murabayashi A.
Tetrahedron, 55(34), 10271-10282 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service