Skip to Content
Merck
All Photos(1)

Key Documents

76523

Supelco

L-Glutamine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Beilstein:
1723797
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

185 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1

InChI key

ZDXPYRJPNDTMRX-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-glutamine is found to be one of the most abundant amino acid in blood stream. It is useful in a number of biochemical processes like production of other amino acids, glucose, nucleotides, protein, and glutathione. It may also be used as a supplement to increase the intestinal villous height, maintain mucosal integrity and stimulate gut mucosal cellular proliferation. It may also find applications in the treatment of chemotherapy and radiotherapy, since it has the tendency to suppress gut and oral-toxic side effects.
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

L-Glutamine is an essential amino acid that is a crucial component of culture media that serves as a major energy source for cells in culture. L-Glutamine is very stable as a dry powder and as a frozen solution. In liquid media or stock solutions, however, L-glutamine degrades relatively rapidly. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therapeutic considerations of L-glutamine: a review of the literature
LAM
Alternative Medicine Review, 4, 239-248 (1999)
Misty R Jenkins et al.
The Journal of experimental medicine, 212(3), 307-317 (2015-03-04)
Failure of cytotoxic T lymphocytes (CTLs) or natural killer (NK) cells to kill target cells by perforin (Prf)/granzyme (Gzm)-induced apoptosis causes severe immune dysregulation. In familial hemophagocytic lymphohistiocytosis, Prf-deficient infants suffer a fatal "cytokine storm" resulting from macrophage overactivation, but
Lisa A Reynolds et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2984-2993 (2014-08-13)
Helminth parasites remain one of the most common causes of infections worldwide, yet little is still known about the immune signaling pathways that control their expulsion. C57BL/6 mice are chronically susceptible to infection with the gastrointestinal helminth parasite Heligmosomoides polygyrus.
Kohichi Matsunaga et al.
The Journal of cell biology, 190(4), 511-521 (2010-08-18)
Autophagy is a catabolic process that allows cells to digest their cytoplasmic constituents via autophagosome formation and lysosomal degradation. Recently, an autophagy-specific phosphatidylinositol 3-kinase (PI3-kinase) complex, consisting of hVps34, hVps15, Beclin-1, and Atg14L, has been identified in mammalian cells. Atg14L
Frédéric Perros et al.
Journal of the American College of Cardiology, 65(7), 668-680 (2015-02-14)
Endothelial cell (EC) dysfunction plays a central role in the pathogenesis of pulmonary arterial hypertension (PAH), promoting vasoconstriction, smooth muscle proliferation, and inflammation. This study sought to test the hypothesis that nebivolol, a β1-antagonist and β2,3-agonist, may improve PAH and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service