Skip to Content
Merck
All Photos(2)

Key Documents

72590

Supelco

Nitrite ionophore I

Selectophore, function tested

Synonym(s):

Cyanoaqua-cobyrinic acid heptakis(2-phenylethyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C102H117ClCoN5O19
CAS Number:
Molecular Weight:
1811.43
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

crystals

quality

function tested

storage temp.

−20°C

SMILES string

[H]O[H].[Co+]C#N.[O-]Cl(=O)(=O)=O.CC1=C2NC(=CC3=NC([C@@H](CCC(=O)OCCc4ccccc4)C3(C)C)=C(C)C5=NC([C@H](CC(=O)OCCc6ccccc6)[C@@]5(C)CCC(=O)OCCc7ccccc7)[C@]8(C)N=C1[C@@H](CCC(=O)OCCc9ccccc9)[C@]8(C)CC(=O)OCCc%10ccccc%10)[C@@H](CCC(=O)OCCc%11ccccc%11)[C@]2(C)CC(=O)OCCc%12ccccc%12

InChI

1S/C101H116N4O14.CN.ClHO4.Co.H2O/c1-70-92-80(45-48-86(107)114-60-52-73-32-18-10-19-33-73)97(3,4)84(103-92)67-83-79(44-47-85(106)113-59-51-72-30-16-9-17-31-72)99(6,68-90(111)118-64-56-77-40-26-14-27-41-77)95(102-83)71(2)93-81(46-49-87(108)115-61-53-74-34-20-11-21-35-74)100(7,69-91(112)119-65-57-78-42-28-15-29-43-78)101(8,105-93)96-82(66-89(110)117-63-55-76-38-24-13-25-39-76)98(5,94(70)104-96)58-50-88(109)116-62-54-75-36-22-12-23-37-75;1-2;2-1(3,4)5;;/h9-43,67,79-82,96,102H,44-66,68-69H2,1-8H3;;(H,2,3,4,5);;1H2/q;;;+1;/p-1/b83-67-,92-70-,95-71-;;;;/t79-,80-,81-,82+,96?,98-,99+,100+,101+;;;;/m1..../s1

InChI key

QRDDGUVPZIMYBN-JXHUONGFSA-M

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore for nitrite-selective liquid membrane electrodes[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Stepanek et al.
Analytica Chimica Acta, 182, 83-83 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service