Skip to Content
Merck
All Photos(1)

Key Documents

52096

Supelco

[(3R)-3-Hydroxyhexadecanoyl]-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-2-[[(3R)-3-hydroxy-1-oxohexadecyl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H45NO5
CAS Number:
Molecular Weight:
415.61
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Assay

≥95.0% (TLC)

optical activity

[α]/D -20.5±3.5°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

C[N+](C)(C)C[C@H](OC(C[C@H](O)CCCCCCCCCCCCC)=O)CC([O-])=O

InChI key

XKAZIAFZAQAHHG-NHCUHLMSSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service