Skip to Content
Merck
All Photos(2)

Key Documents

46279

Supelco

Cyanophos

PESTANAL®, analytical standard

Synonym(s):

O-(4-Cyanophenyl) O,O-dimethyl phosphorothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10NO3PS
CAS Number:
Molecular Weight:
243.22
Beilstein:
2695901
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(cc1)C#N

InChI

1S/C9H10NO3PS/c1-11-14(15,12-2)13-9-5-3-8(7-10)4-6-9/h3-6H,1-2H3

InChI key

SCKHCCSZFPSHGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyanophos is an organophosphate insecticide, which is effectively used against various root-parasitic nematodes such as Diaspididae, Lepidoptera, Coccidae, Aphididae and other insects on various fruits and vegetables.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Cyanophos may be used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography coupled to mass spectrometry (LC-MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid simultaneous determination for organophosphorus
pesticides in human serum by LC-MS
Inoue S, et al.
Journal of Pharmaceutical and Biomedical Analysis, 44(2), 258-264 (2007)
Enhancing agents for phytoremediation of soil contaminated by cyanophos
Romeh AA
Ecotoxicology and Environmental Safety, 117(2), 124-131 (2015)
W C Mullié et al.
Ecotoxicology and environmental safety, 43(1), 1-10 (1999-05-20)
Seven roosts of red-billed quelea, Quelea quelea, in the Senegal River Valley and Delta were visited during and after aerial or terrestrial treatments with either Cyanox (cyanophos 500 g a.i. liter-1, five roosts) or Queletox (fenthion 640 g a.i. liter-1
[Evaluation of bird poison residues in "millet-eaters" (Quelea quelea L.). Toxicologic consequences arising from the consumption of poisoned birds by domestic animals and man. 2. Cyanophos].
B S Cisse et al.
Dakar medical, 29(2), 377-382 (1984-01-01)
[Contamination of sugar cane by 2 bird poisons: fenthion and cyanophos. Evaluation of residues in sugar cane juice transformed into sugar].
B S Cisse et al.
Dakar medical, 29(2), 383-390 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service