Skip to Content
Merck
All Photos(2)

Key Documents

45305

Supelco

Dicrotophos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16NO5P
CAS Number:
Molecular Weight:
237.19
Beilstein:
1880084
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=O)(OC)O\C(C)=C\C(=O)N(C)C

InChI

1S/C8H16NO5P/c1-7(6-8(10)9(2)3)14-15(11,12-4)13-5/h6H,1-5H3/b7-6+

InChI key

VEENJGZXVHKXNB-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dietrich E Lorke et al.
Current pharmaceutical design, 23(23), 3432-3439 (2016-11-02)
Reversible cholinesterase inhibitors, when given prophylactically before exposure to organophosphates, are able to decrease organophosphate-induced mortality. However, the efficacy of pyridostigmine, the only pre-treatment substance approved by the US Federal Drug Administration, is unsatisfactory. In search of a better prophylactic
Moneen M Jones et al.
Journal of economic entomology, 111(2), 829-835 (2018-01-24)
To assess the toxicity of bifenthrin and four mixtures of insecticides to tarnished plant bug, we used an insecticide dip method of green bean to treat adults of a laboratory colony; mortality was assessed after 48 h. LC50s for imidacloprid
E L Flickinger et al.
Journal - Association of Official Analytical Chemists, 67(4), 827-828 (1984-07-01)
About 1100 birds of 12 species died from organophosphate poisoning in Matagorda County on the Texas Gulf Coast in March and May 1982. Birds died from feeding on rice seed that was illegally treated with dicrotophos or monocrotophos and placed
A L Kilpatrick et al.
Journal of economic entomology, 98(3), 814-820 (2005-07-19)
Certain neonicotinoids are used in cotton, Gossypium hirsutum (L.), to control various piercing-sucking pests. We conducted field studies using three neonicotinoids (acetamiprid, thiamethoxam, and imidacloprid) and an organophosphate (dicrotophos) to assess the activity of these insecticides against nontarget arthropods, particularly
R Meiniel
Teratology, 23(2), 259-271 (1981-04-01)
A comparative qualitative and quantitative study of the embryotoxic and teratogenic activities of various chemical agents known to have neuromuscular blocking properties was done on the quail embryo. Intrinsic embryotoxic activity and teratogenicity were evaluated for each agent, and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service