Skip to Content
Merck
All Photos(1)

Key Documents

44920

Sigma-Aldrich

Iron(III) acetylacetonate

purum, ≥97.0% (RT)

Synonym(s):

2,4-Pentanedione iron(III) derivative, Fe(acac)3, Ferric acetylacetonate, Iron(III) 2,4-pentanedionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(C5H7O2)3
CAS Number:
Molecular Weight:
353.17
Beilstein:
4157960
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥97.0% (RT)

form

powder

reaction suitability

core: iron
reagent type: catalyst

mp

180-182 °C (dec.) (lit.)
180-190 °C (dec.)

density

5.24 g/mL at 25 °C (lit.)

SMILES string

CC(=O)\C=C(\C)O[Fe](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.Fe/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3/b3*4-3-;

InChI key

AQBLLJNPHDIAPN-LNTINUHCSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) acetylacetonate, also known as tris(acetylacetonato)iron(III) or Fe(acac)3, is a coordination compound that is highly soluble in organic solvents. It is used as a precursor in the preparation of iron-containing materials and thin films, which are important in electronics and coatings. Additionally, it is also as a catalyst in organic reactions, particularly in the synthesis of organic compounds and polymerization processes.

Application

Iron(III) acetylacetonate can be used as:     
  • A precursor in the synthesis of Fe3O4/carbon composite fibers via forcespinning. This fiber material is suitable for use as anode materials in lithium-ion batteries.     
  • A precursor in the synthesis of iron-based metal-organic frameworks (MOFs) for the applications in rechargeable alkali-ion batteries.
  • A catalyst in the controlled living radical polymerization of vinyl acetate to synthesis of polymers with controlled molecular weights.   
  • A precursor to synthesize iron-doped titania (Fe-TiO2) photocatalysts for the potential applications in photocatalytic reactions.     
  • A MOCVD precursor for highly crystalline (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.

MOCVD precursor for highly crystalline (Zn,Fe)Fe2O4 films and magnetic property measurements of these films.

Other Notes

Catalyst used in Grignard reactions with acid chlorides to give ketones[1][2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Cardellicchio et al.
Tetrahedron Letters, 28, 2053-2053 (1987)
V. Fiandanese et al.
Tetrahedron Letters, 25, 4805-4805 (1984)
Tingting Li et al.
Nanoscale, 7(9), 4171-4178 (2015-02-12)
The excellent electrochemical performance of greigite (Fe3S4) coupled with its vast abundance and low toxicity make it a good prospect as an anode material for lithium ion batteries (LIBs). In this research, a simple and feasible approach for producing pure
Roland Schoch et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 15(17), 3768-3775 (2014-09-13)
A new iron-based catalyst for carbon monoxide oxidation, as a potential substitute for precious-metal systems, has been prepared by using a facile impregnation method with iron tris-acetylacetonate as a precursor on γ-Al2 O3 . Light-off and full conversion temperatures as
Michael Challenor et al.
Dalton transactions (Cambridge, England : 2003), 43(44), 16780-16787 (2014-10-07)
We report the synthesis, characterisation and evaluation of the in vitro biocompatibility of polymeric nanoparticles with both magnetic and upconverting fluorescent properties. The particles consist of superparamagnetic iron oxide nanoparticles and upconverting NaYF4:Yb,Er nanoparticles co-encapsulated within a poly(glycidyl methacrylate) sphere.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service