Skip to Content
Merck
All Photos(1)

Documents

44028

Supelco

Cyclohexene

analytical standard, ≥99.5% (GC)

Synonym(s):

Tetrahydrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10
CAS Number:
Molecular Weight:
82.14
Beilstein:
906737
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.8 (vs air)

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

590 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.01% 2,6-di-tert-butyl-4-methylphenol as stabilizer

expl. lim.

5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.446 (lit.)
n20/D 1.446

bp

83 °C (lit.)

mp

−104 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

C1CCC=CC1

InChI

1S/C6H10/c1-2-4-6-5-3-1/h1-2H,3-6H2

InChI key

HGCIXCUEYOPUTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelia W Ray et al.
The journal of physical chemistry. A, 116(25), 6720-6730 (2012-05-29)
Earlier synchrotron photoionization mass spectrometry experiments suggested a prominent ring-opening channel in the OH-initiated oxidation of cyclohexene, based on comparison of product photoionization spectra with calculated spectra of possible isomers. The present work re-examines the OH + cyclohexene reaction, measuring
Jan Lennard Wolf et al.
Physical chemistry chemical physics : PCCP, 13(23), 10952-10964 (2011-03-29)
The gas-phase ozonolysis of cyclic-alkenes (1-methyl-cyclohexene, methylene-cyclohexane, α-pinene, β-pinene) is studied with respect to the pressure dependent formation of secondary organic aerosol (SOA). We find that SOA formation is substantially suppressed at lower pressures for all alkenes under study. The
Oleg Y Lyakin et al.
Inorganic chemistry, 50(12), 5526-5538 (2011-05-24)
Complexes [(BPMEN)Fe(II)(CH(3)CN)(2)](ClO(4))(2) (1, BPMEN = N,N'-dimethyl-N,N'-bis(2-pyridylmethyl)-1,2-diaminoethane) and [(TPA)Fe(II)(CH(3)CN)(2)](ClO(4))(2) (2, TPA = tris(2-pyridylmethyl)amine) are among the best nonheme iron-based catalysts for bioinspired oxidation of hydrocarbons. Using EPR and (1)H and (2)H NMR spectroscopy, the iron-oxygen intermediates formed in the catalyst systems
Emanuele Monza et al.
The journal of physical chemistry. A, 115(45), 12864-12878 (2011-07-19)
The source function (SF) introduced in late 90s by Bader and Gatti quantifies the influence of each atom in a system in determining the amount of electron density at a given point, regardless of the atom's remote or close location
Oxidative decarboxylation of benzilic acid by a biomimetic iron(II) complex: evidence for an iron(IV)-oxo-hydroxo oxidant from O2.
Sayantan Paria et al.
Angewandte Chemie (International ed. in English), 50(47), 11129-11132 (2011-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service