Skip to Content
Merck
All Photos(1)

Key Documents

436674

Supelco

Amberlite FPA66 Ion Exchange Resin

free base, 16-50 mesh, anionic

Synonym(s):

strong cation exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
SB.52

Product Name

AmberLite FPA66 Anion Exchange Resin free base,

form

beads

Quality Level

parameter

60 °C OH- form max. temp.

moisture

40-46%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroporous)

matrix active group

polyamine functional group

particle size

300-1200 μm
50-125 mesh

operating pH

0-7

capacity

1.35 meq/mL by wetted bed volume

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

This resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.
Specifications:

Application

AmberLite FPA66 Anion Exchnage Resin free base was used to prepare a catalyst during the production of myrtanal.
Weakly basic anion exchange resin for deashing and mixed bed polishing of high fructose corn syrups. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformation of Biomass Products into Fine Chemicals Catalyzed by Solid Lewis-and Br?nsted-acids.
Corma, Avelino, Michael Renz, and Manuel Susarte.
Topics in Catalysis, 52, 1182-1189 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service