Skip to Content
Merck
All Photos(1)

Documents

428698

Supelco

Ambersep (formerly Dowex) Retardion 11A8 Ion Exchange Resin

50-100 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

product name

AmberSep® (formerly Dowex) Retardion 11A8, 50-100 mesh

description

amphoteric form

Quality Level

form

beads

parameter

70 °C max. temp.

moisture

43-48%

technique(s)

LPLC: suitable

color

deep yellow, tan

matrix

amphoteric styrene-divinylbenzene (gel)

particle size

50-100 mesh

operating pH

0-14

capacity

0.7 meq/mL by wetted bed volume

separation technique

mixed mode

Looking for similar products? Visit Product Comparison Guide

General description

Amphoteric properties of Dowex Retardion 11A8 allows specific isolation of elements from complex mixtures which generally is possible with use of monofunctional ion exchangers. Determination of trace amounts of Zinc and Cadmium by radiochemical NAA was performed using Retardion 11A8.
The Retardion 11A8 is not a true mixed bed resin, but a resin with mixed functionality. It contains paired anion and cation exchange sites (each exists as the other′s counter-ion) that adsorb mobile ions from a bulk stream.

Application

Dowex Retardion 11A8 was used in solid phase extraction for determination of total concentrations by flame atomic absorption spectrometry.
Chromatographic resin for ion separation. Useful for removal of ionic detergents (e.g., SDS) from protein samples. It is used for the removal of electrolytes from water solutions, and separation of cations from anions. Desalting occurs by SEC-type mechanism, so column use is recommended for best success.
     

Physical form

Mixed bed

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Ambersep is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion Exchange Behaviour of Cadmium on Amphoteric Ion Exchange Resin Retardion lIAS and its Application for the Determination of Cadmium in Biological Materials by Neutron Activation Analysis.
Samczynski, Zbigniew, and Rajmund Dybczynski.
Chem. Anal. (Warsaw), 41, 873-873 (1996)
Pre-concentration of traces of cadmium, cobalt, nickel and lead in natural honeys by solid phase extraction followed by their determination using flame atomic absorption spectrometry.
Stecka, Helena, et al.
Journal of the Brazilian Chemical Society, 25, 331-339 (2014)
The use of the amphoteric ion-exchange resin retardion 11A8 for inorganic separations.
Dybczynski, Rajmund, and Elzbieta Sterlinska.
Journal of Chromatography A, 102, 263-271 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service