Skip to Content
Merck
All Photos(3)

Key Documents

390062

Sigma-Aldrich

Thiazole Orange

Dye content ~90 %

Synonym(s):

1-Methyl-4-[(3-methyl-2(3H)-benzothiazolylidene)methyl]quinolinium p-tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24N2O3S2
CAS Number:
Molecular Weight:
476.61
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

powder

Quality Level

composition

Dye content, ~90%

technique(s)

titration: suitable

mp

270 °C (dec.) (lit.)

λmax

512 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.CN2C(\Sc3ccccc23)=C\c4cc[n+](C)c5ccccc45

InChI

1S/C19H17N2S.C7H8O3S/c1-20-12-11-14(15-7-3-4-8-16(15)20)13-19-21(2)17-9-5-6-10-18(17)22-19;1-6-2-4-7(5-3-6)11(8,9)10/h3-13H,1-2H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

ACOJCCLIDPZYJC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Thiazole Orange can be tethered with peptide nucleic acid for the detection of target nucleic acid in homogeneous solution.[1] It is a dye with, both predicted physical characteristics and the biological staining characteristics of a good reticulocyte dye, hence it is considered as a dye for reticulocyte analysis. It has massive fluorescence enhancement and high quantum yield upon binding to nucleic acids, particularly RNA. It is non precipitating and can permeates through live cell membranes.[2]

Application

Thiazole orange has been used for the staining and thereby quantification of reticulocytes.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam D Cawte et al.
Nature communications, 11(1), 1283-1283 (2020-03-11)
RNA molecules play vital roles in many cellular processes. Visualising their dynamics in live cells at single-molecule resolution is essential to elucidate their role in RNA metabolism. RNA aptamers, such as Spinach and Mango, have recently emerged as a powerful
Danitza Nébor et al.
Scientific reports, 8(1), 12793-12793 (2018-08-26)
Anemic Nan mice carry a mutation (E339D) in the second zinc finger of erythroid transcription factor KLF1. Nan-KLF1 fails to bind a subset of normal KLF1 targets and ectopically binds a large set of genes not normally engaged by KLF1
Eeman Khorramian et al.
Shock (Augusta, Ga.), 48(4), 490-497 (2017-04-30)
Intensive care unit (ICU) anemia is an extreme version of anemia of inflammation that occurs commonly in critically ill patients and is associated with increased morbidity and mortality. Currently available therapies for ICU anemia have shown inconsistent efficacies in clinical
Light-up probes: thiazole orange-conjugated peptide nucleic acid for detection of target nucleic acid in homogeneous solution.
Svanvik N
Analytical Biochemistry, 281(1), 26-35 (2000)
Costimulation blockade, busulfan, and bone marrow promote titratable macrochimerism, induce transplantation tolerance, and correct genetic hemoglobinopathies with minimal myelosuppression.
Adams AB
Journal of Immunology, 167(2), 1103-1111 (2001)

Questions

  1. How can i make a solution or How is the concentration of thiazole orange??

    1 answer
    1. This product is not tested for solubility. For absorbance testing, it is dissolved in methanol. Reference papers describe preparing stock solutions at 1mg/ml in methanol. There are 10 mM solutions available from other suppliers that are prepared in DMSO.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service