Skip to Content
Merck
All Photos(1)

Key Documents

34334

Supelco

2,4-Dinitrophenol

PESTANAL®, analytical standard

Synonym(s):

α-Dinitrophenol, 2,4-DNP, DNP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3OH
CAS Number:
Molecular Weight:
184.11
Beilstein:
1246142
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

6.35 (vs air)

product line

PESTANAL®

quality

(with approx. 0,3 ml H2O/g)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

108-112 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H

InChI key

UFBJCMHMOXMLKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitrophenol is called as an uncoupler of oxidative phosphorylation, which can prevent adenosine triphosphate(ATP) synthesis, but it permits electron transfer to proceed.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Desen. Expl. 4 - STOT RE 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemistry Biochemistry: Solutions Manual (1998)
Biochemistry Biochemistry: Solutions Manual (1998)
Stephanie C Weber et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(19), 7338-7343 (2012-04-21)
Chromosomal loci jiggle in place between segregation events in prokaryotic cells and during interphase in eukaryotic nuclei. This motion seems random and is often attributed to brownian motion. However, we show here that locus dynamics in live bacteria and yeast
Tatsuya Oka et al.
The Journal of allergy and clinical immunology, 132(4), 922-932 (2013-07-03)
Rapid desensitization transiently prevents severe allergic reactions, allowing administration of life-saving therapies in previously sensitized patients. However, the mechanisms underlying successful rapid desensitization are not fully understood. We sought to investigate whether the mast cell (MC) is an important target
Toni-Maree Rogers et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 10(4), 611-618 (2015-03-20)
The use of targeted therapies toward specific oncogenic driver mutations has become a critical factor in the treatment of patients with lung cancer. It is therefore essential to utilize tests with high performance characteristics. Fluorescence in situ hybridization (FISH) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service