Skip to Content
Merck
All Photos(1)

Key Documents

33371

Supelco

Naptalam

PESTANAL®, analytical standard

Synonym(s):

N-(1-Naphthyl)phthalamidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13NO3
CAS Number:
Molecular Weight:
291.30
Beilstein:
2814102
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

185-190 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1ccccc1C(=O)Nc2cccc3ccccc23

InChI

1S/C18H13NO3/c20-17(14-9-3-4-10-15(14)18(21)22)19-16-11-5-7-12-6-1-2-8-13(12)16/h1-11H,(H,19,20)(H,21,22)

InChI key

JXTHEWSKYLZVJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naptalam is a purple crystalline solid, which can be used as an anti-geotropic agent and as an auxin (IAA) antagonist in plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monaco, JT, et al.
Weed Sci. (2002)
Hyo-Jun Lee et al.
The New phytologist, 225(3), 1285-1296 (2019-07-25)
Plants sense mechanical stimuli to recognise nearby obstacles and change their growth patterns to adapt to the surrounding environment. When roots encounter an obstacle, they rapidly bend away from the impenetrable surface and find the edge of the barrier. However
Naiyanate Jaroensanti et al.
Bioscience, biotechnology, and biochemistry, 78(11), 1839-1849 (2014-10-30)
Brassinosteroid (BR) and auxin co-regulate plant growth in a process termed cross-talking. Based on the assumption that their signal transductions are partially shared, inhibitory chemicals for both signal transductions were screened from a commercially available library. A chemical designated as
Richard J Pattison et al.
The Plant journal : for cell and molecular biology, 70(4), 585-598 (2012-01-04)
The temporal and spatial control of auxin distribution has a key role in the regulation of plant growth and development, and much has been learnt about the mechanisms that influence auxin pools and gradients in vegetative tissues, particularly in Arabidopsis.
Yingnan Chen et al.
Plant biotechnology journal, 10(2), 139-149 (2011-07-23)
Crop architecture parameters such as tiller number, angle and plant height are important agronomic traits that have been considered for breeding programmes. Auxin distribution within the plant has long been recognized to alter architecture. The rice (Oryza sativa L.) genome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service