Skip to Content
Merck
All Photos(1)

Documents

32805

Supelco

Mandipropamid

PESTANAL®, analytical standard

Synonym(s):

2-(4-Chlorophenyl)-N-[3-methoxy-4-(2-propynyloxy)phenethyl]-2-(2-propynyloxy)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClNO4
CAS Number:
Molecular Weight:
411.88
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=CC=C(C(OCC#C)C(NCCC2=CC=C(OCC#C)C(OC)=C2)=O)C=C1

InChI

1S/C23H22ClNO4/c1-4-14-28-20-11-6-17(16-21(20)27-3)12-13-25-23(26)22(29-15-5-2)18-7-9-19(24)10-8-18/h1-2,6-11,16,22H,12-15H2,3H3,(H,25,26)

InChI key

KWLVWJPJKJMCSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mandipropamid is a carboxylic acid amide, which belongs to the oomycete class of fungicides. It is found to be effective in controlling phytopathogens such as Plasmopara viticola on grapes and potato late blight caused by Phytophthora infestans.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Guofeng Xu et al.
Environmental science and pollution research international, 27(32), 40148-40155 (2020-07-15)
Enantioselective monitoring of chiral fungicide mandipropamid enantiomers were carried out in grapes and wine-making process. The enantiomers of mandipropamid were separated on a Lux Cellulose-2 column and determined by ultra-high-performance liquid chromatography coupled with tandem mass spectrometry (UHPLC-MS/MS). The processing
Mandipropamid targets the cellulose synthase?like PiCesA3 to inhibit cell wall biosynthesis in the oomycete plant pathogen, Phytophthora infestans.
Blum M, et al.
Molecular Plant Pathology, 11(2), 227-243 (2010)
Multicomponent reactions in fungicide research: The discovery of mandipropamid.
Lamberth C, et al.
Bioorganic & Medicinal Chemistry, 16(3), 1531-1545 (2008)
Vasileios Vangalis et al.
Journal of fungi (Basel, Switzerland), 6(4) (2020-12-11)
Woronin bodies are membrane-bound organelles of filamentous ascomycetes that mediate hyphal compartmentalization by plugging septal pores upon hyphal damage. Their major component is the peroxisomal protein Hex1, which has also been implicated in additional cellular processes in fungi. Here, we
Nikhilesh Dhar et al.
Plant disease, 104(1), 82-93 (2019-11-19)
The lettuce downy mildew pathogen, Bremia lactucae, is an obligate oomycete that causes extensive produce losses. Initial chlorotic symptoms that severely reduce the market value of the produce are followed by the appearance of white, downy sporulation on the abaxial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service