Skip to Content
Merck
All Photos(3)

Key Documents

306975

Sigma-Aldrich

tert-Butyl methyl ether

anhydrous, 99.8%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.8%
bp:
55-56 °C (lit.)
vapor pressure:
4.05 psi
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.1 (vs air)

vapor pressure

4.05 psi

Assay

99.8%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

mp

-110 °C

density

0.74 g/mL at 25 °C (lit.)

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl methyl ether (MTBE) is a gasoline additive. MTBE undergoes oxidative degradation in the presence of propane-oxidizing bacterial strains.[1] The kinetic studies of heat-assisted persulfate oxidation of MTBE under various parameters suggests that the reaction follows the pseudo-first-order kinetics.[2] MTBE can be synthesized by acid catalyzed reaction between methanol and isobutene.[3] A study suggests that the addition of MTBE increases the number of active sites during polymerization of propene by stopped-flow method.[4]
tert-Butyl methyl ether is a commonly used organic solvent that can be synthesized by acid catalyzed reaction between methanol and isobutene.[5] It is an effective alternative to lead containing additives for enhancing the octane rating of gasoline.[6]

Application

tert-Butyl methyl ether may be used to synthesize fatty acid methyl esters (FAMEs) and glycerol tert-butyl ether via transesterification with canola oil under supercritical conditions.[7]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

<BIG>Eagleson M. </BIG>
Concise Encyclopedia Chemistry, 154-154 (1995)
Application of artificial neural networks for modeling of the treatment of wastewater contaminated with methyl tert-butyl ether (MTBE) by UV/H 2 O 2 process.
Salari D, et al.
Journal of Hazardous Materials, 125(1), 205-210 (2005)
Viktor Nilsson et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(11), 1166-1176 (2020-04-21)
To elucidate what properties control and practically limit ion transport in highly concentrated electrolytes (HCEs), the viscosity, ionic conductivity, ionicity, and transport numbers were studied for nine model electrolytes and connected to the rate capability in Li-ion battery (LIB) cells.
Relative solubility, stability, and absorptivity of lutein and β-carotene in organic solvents.
Craft NE and Soares JH.
Journal of Agricultural and Food Chemistry, 40(3), 431-434 (1992)
New approach of catalyst-free biodiesel production from canola oil in supercritical tert-butyl methyl ether (MTBE).
Farobie O, et al.
Fuel: The Science and Technology of Fuel and Energy, 135, 172-181 (2014)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How is the dehydration process for this product? Is it through molecular sieves or by using sodium?

    1 answer
    1. Molecular sieves are employed to dehydrate the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service