Skip to Content
Merck
All Photos(2)

Key Documents

230308

Sigma-Aldrich

1,3-Diphenylacetone p-tosylhydrazone

98%

Synonym(s):

1,3-Diphenyl-2-propanone p-tosylhydrazone, 1,3-Diphenyl-2-propanone tosylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NHN=C(CH2C6H5)2
CAS Number:
Molecular Weight:
378.49
Beilstein:
2225293
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Assay

98%

form

powder or crystals

mp

187-188 °C (lit.)

solubility

chloroform: 25 mg/mL, clear, orange

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1ccc(cc1)S(=O)(=O)N\N=C(\Cc2ccccc2)Cc3ccccc3

InChI

1S/C22H22N2O2S/c1-18-12-14-22(15-13-18)27(25,26)24-23-21(16-19-8-4-2-5-9-19)17-20-10-6-3-7-11-20/h2-15,24H,16-17H2,1H3

InChI key

GDXUEUWCUUAZFM-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service