Skip to Content
Merck
All Photos(1)

Key Documents

08578

Sigma-Aldrich

2-Amino-2-methyl-1-propanol

BioUltra, ≥99.0% (GC)

Synonym(s):

β-Aminoisobutyl alcohol, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)CH2OH
CAS Number:
Molecular Weight:
89.14
Beilstein:
505979
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

vapor density

3 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 25 °C)

product line

BioUltra

Assay

≥99.0% (GC)

form

solid

impurities

insoluble matter, passes filter test

refractive index

n20/D 1.4455 (lit.)
n20/D 1.448

pH

11.0-12.0 (25 °C, 0.1 M in H2O)

useful pH range

9.0-10.5

pKa (25 °C)

9.7

bp

165 °C (lit.)

mp

24-28 °C (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

density

0.934 g/mL at 25 °C (lit.)

anion traces

carbonate (CO32-): ≤1000 mg/kg
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CC(C)(N)CO

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

InChI

1S/C4H11NO/c1-4(2,5)3-6/h6H,3,5H2,1-2H3

InChI key

CBTVGIZVANVGBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Amino-2-methyl-1-propanol can be used to study biological buffers. 2-Amino-2-methyl-1-propanol has been used in a study to report on an ATR-FTIR spectroscopic investigation of the CO(2) absorption characteristics of a series of heterocyclic diamines. 2-Amino-2-methyl-1-propanol has also been used in a study to develop a mild and convenient method for preparing 4,4-dimethyloxazoline (DMOX) derivatives of fatty acids for GC-MS analysis.

Caution

may solidify at room temperature

Other Notes

Determination of alkaline phosphatase; Stabilizes the Calmagite reagent in the automated measurement of magnesium in serum and urine.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

179.8 °F - closed cup

Flash Point(C)

82.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H J Derks et al.
Clinical chemistry, 27(2), 318-321 (1981-02-01)
When 2-amino-2-methyl-1-propanol buffer solutions are extracted with chloroform, impurities inhibiting alkaline phosphatase are selectively removed and this source of variation is thereby greatly decreased. We also studied the effect of adding zinc sulfate to this buffer. A combination of chloroform
Kelly Robinson et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(9), 2331-2341 (2012-04-21)
This paper reports on an ATR-FTIR spectroscopic investigation of the CO(2) absorption characteristics of a series of heterocyclic diamines: hexahydropyrimidine (HHPY), 2-methyl and 2,2-dimethylhexahydropyrimidine (MHHPY and DMHHPY), hexahydropyridazine (HHPZ), piperazine (PZ) and 2,5- and 2,6-dimethylpiperazine (2,6-DMPZ and 2,5-DMPZ). By using
J.-P. Bretaudiere and T. Spillman et al.
Methods of Enzymatic Analysis, 4, 75-75 (1984)
Stabilization of the calmagite reagent for automated measurement of magnesium in serum and urine.
J Savory et al.
Clinical chemistry, 31(3), 487-488 (1985-03-01)
M Lamberto et al.
Lipids, 29(6), 441-444 (1994-06-01)
The structures of two unusual fatty acids, the known trans-3-hexadecenoic acid and a novel trans-3-tetradecenoic acid, both isolated from the Nova Scotian seaweeds Palmaria palmata and Chondrus crispus, were positively identified. After the extraction of the total fatty acids by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service