Skip to Content
Merck
All Photos(1)

Key Documents

05832

Supelco

all-cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester

analytical standard

Synonym(s):

Fame 22:6n-3, Methyl DHA, Methyl all-cis-4,7,10,13,16,19-docosahexenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O2
CAS Number:
Molecular Weight:
342.51
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

format

neat

functional group

ester

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(=O)OC

InChI

1S/C23H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h4-5,7-8,10-11,13-14,16-17,19-20H,3,6,9,12,15,18,21-22H2,1-2H3/b5-4-,8-7-,11-10-,14-13-,17-16-,20-19-

InChI key

VCDLWFYODNTQOT-JDPCYWKWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

all-cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester belongs to the class of very long-chain fatty acid methyl esters.

Application

This analytical standard can also be used as follows:
  • Development of a gas chromatography method combined with a quadrupole time-of-flight mass analyzer (GC-QTOF) functioning with an atmospheric pressure chemical ionization (APCI) source for the detection of 11 methyl esters of very long chain-polyunsaturated fatty acids (VLU-PUFAs) in 12 eyes samples of four different fish species
  • Gas chromatography-mass spectrometric (GC-MS) analysis of eicosapentaenoic acid and docosahexaenoic acids, separated from fish oil samples by pH-zone-refining countercurrent chromatography following their methyl esterification
  • Quantification of eicosapentaenoic acid and docosahexaenoic acids following their ultrasonic cell crusher extraction and derivatization to methyl esters from six types of seafood samples by gas chromatography coupled to flame ionization detector (GC-FID)
  • Optimization of a capillary zone electrophoresis procedure coupled with direct absorption UV detection (CZE-UV) to determine eicosapentaenoic acid and docosahexaenoic acids after their methylation in 10 different samples of commercial omega-3 supplements
  • Simultaneous analysis of 45 fatty acids following their methyl derivatization in human blood cells by gas chromatography-tandem mass spectrometry (GC-MS/MS) with an ammonia-induced chemical ionization (CI)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what is the solubility of this DHA methyl ester standard? It is immiscible in n-hexane.

    1 answer
    1. This product is not tested for solubility on a lot to lot basis. For assay purposes, this material has been solubilized at 10 mg/mL in Heptane and n-Hexane. Historical data indicates that this compound is also soluble at 50 mg/mL in Chloroform. This is a neat or pure liquid with a density of 0.921 g/mL at 25 °C.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service