Skip to Content
Merck
All Photos(1)

Documents

03762

Sigma-Aldrich

2,6-Di-tert-butyl-4-methylphenol

tested according to Ph. Eur.

Synonym(s):

Butylhydroxytoluenum, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

autoignition temp.

878 °F

bp

265 °C (lit.)

mp

69-73 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application


  • Quantitative analysis of residual butylated hydroxytoluene and butylated hydroxyanisole in Salmo salar, milk, and butter by liquid chromatography-tandem mass spectrometry.: This study provides a quantitative analysis of residual 2,6-Di-tert-butyl-4-methylphenol (BHT) and butylated hydroxyanisole (BHA) in various food products. The research highlights the presence and concentration of these antioxidants in fish, milk, and butter, demonstrating their widespread use in food preservation (Galal et al., 2024).

  • The key factors of solid nanodispersion for promoting the bioactivity of abamectin.: This research investigates the use of BHT in solid nanodispersions to enhance the bioactivity of abamectin, a widely used insecticide. The study identifies key factors that improve the stability and effectiveness of the formulation, suggesting applications in agricultural biotechnology (Ding et al., 2024).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Li et al.
Circulation, 105(7), 861-867 (2002-02-21)
Current epidemiological data suggest that dental infections affecting tooth-supporting tissues (periodontitis) can disseminate into the systemic circulation and thereby contribute to atherosclerosis progression. To test this hypothesis, we investigated the effect of repeated systemic inoculations with Porphyromonas gingivalis (Pg), a
I Björkhem et al.
Arteriosclerosis and thrombosis : a journal of vascular biology, 11(1), 15-22 (1991-01-01)
Rabbits fed a 1% cholesterol diet with or without the antioxidant butylated hydroxytoluene (BHT) developed typical atherosclerotic lesions. The addition of BHT gave higher levels of total cholesterol (+40%), triglycerides (+250%), low density lipoprotein (LDL), and very low density lipoprotein
A M Vijesh et al.
European journal of medicinal chemistry, 46(11), 5591-5597 (2011-10-05)
In the present study two new series of Hantzsch 1,4-dihydropyridine derivatives (1,4-DHPs) containing substituted pyrazole moiety (4a-f and 5a-f) were synthesized by the reaction of 3-aryl-1H-pyrazole-4-carbaldehydes with 1,3-dicarbonylcompounds (ethylacetoacetate and methylacetoacetate) and ammonium acetate. The newly synthesized compounds were characterized
Gen-ichiro Arimura et al.
Plant physiology, 146(3), 965-973 (2008-01-01)
Continuous mechanical damage initiates the rhythmic emission of volatiles in lima bean (Phaseolus lunatus) leaves; the emission resembles that induced by herbivore damage. The effect of diurnal versus nocturnal damage on the initiation of plant defense responses was investigated using
Butylated hydroxytoluene (BHT): a review.
H Babich
Environmental research, 29(1), 1-29 (1982-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service