Skip to Content
Merck
All Photos(1)

Key Documents

28-6130

Sigma-Aldrich

Sulfur dioxide solution

SAJ first grade, 5.0% SO2

Synonym(s):

Sulfurous acid, Sulfurous anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO3
CAS Number:
Molecular Weight:
82.08
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
concentration:
5.0% SO2
grade:
SAJ first grade

grade

SAJ first grade

form

liquid

availability

available only in Japan

concentration

5.0% SO2

density

1.03 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

OS(O)=O

InChI

1S/H2O3S/c1-4(2)3/h(H2,1,2,3)

InChI key

LSNNMFCWUKXFEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miquel Pascual et al.
Journal of the science of food and agriculture, 95(15), 3067-3076 (2015-07-16)
Rainfed viticulture, mainly in semi-arid environments, is limited by environmental variability, particularly precipitation and its seasonal distribution, and soil water availability, thus ultimately determining the final quality of grape and wine. Studies on the feasibility of practices such as canopy
Heng Yu et al.
Bioresource technology, 199, 188-193 (2015-09-06)
This study established a new more neutral magnesium bisulfate pretreatment (MBSP) using magnesium bisulfate as sulfonating agent for improving the enzymatic hydrolysis efficiency of corn stover. Using the MBSP with 5.21% magnesium bisulfate, 170°C and pH 5.2 for 60 min

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service