Skip to Content
Merck
All Photos(1)

Documents

24-2885

Sigma-Aldrich

Phthalic acid

SAJ special grade, ≥99.0%

Synonym(s):

1,2-Benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
608199
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

mp

210-211 °C (dec.) (lit.)

SMILES string

OC(C1=C(C(O)=O)C=CC=C1)=O

InChI

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher H Hurst et al.
Toxicological sciences : an official journal of the Society of Toxicology, 74(2), 297-308 (2003-06-14)
Phthalate esters are widely used as plasticizers in the manufacture of products made of polyvinyl chloride. Mono-(2-ethylhexyl)-phthalate (MEHP) induces rodent hepatocarcinogenesis by a mechanism that involves activation of the nuclear transcription factor peroxisome proliferator-activated receptor-alpha (PPARalpha). MEHP also activates PPAR-gamma
Russ Hauser
Seminars in reproductive medicine, 24(3), 156-167 (2006-06-29)
Scientific and public concern about the potential risk of environmental chemicals to male reproductive health has been heightened by reports of downward trends in semen quality, as well as increased rates of developmental urogenital tract anomalies and testicular cancer. Of
Paula J Lapinskas et al.
Toxicology, 207(1), 149-163 (2004-12-14)
Phthalate esters belong to a large class of compounds known as peroxisome proliferators (PP). PP include chemicals that activate different subtypes of the peroxisome proliferator-activated receptor (PPAR) family. The ability of phthalate esters and their metabolites to activate responses through
Maria Cristina Fossi et al.
Marine pollution bulletin, 64(11), 2374-2379 (2012-09-12)
Baleen whales are potentially exposed to micro-litter ingestion as a result of their filter-feeding activity. However, the impacts of microplastics on baleen whales are largely unknown. In this case study of the Mediterranean fin whale (Balaenoptera physalus), we explore the
Juanjuan Feng et al.
Journal of chromatography. A, 1268, 16-21 (2012-11-07)
Because of the occurrence of ion exchange between high-ionic-strength solution and anions of polymeric ionic liquids (PILs), PILs based solid-phase microextraction (SPME) fibers were rarely used in direct immersion mode to high-salt-added samples. In this work, a novel double-confined PIL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service