Skip to Content
Merck
All Photos(1)

Key Documents

21-1430

Sigma-Aldrich

Nicotinic acid

SAJ special grade, ≥99.5%

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.5%

form

powder

availability

available only in Japan

color

white

mp

236-239 °C (lit.)

solubility

ethanol: 12.5 g/L at 25 °C
water: soluble 180 g/L at 20 °C

density

1.473 g/cm3 at 25 °C

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John R Guyton et al.
Journal of the American College of Cardiology, 62(17), 1580-1584 (2013-08-07)
This study sought to examine the relationship between niacin treatment, lipoproteins, and cardiovascular (CV) outcomes in this secondary analysis of the AIM-HIGH (Atherothrombosis Intervention in Metabolic Syndrome With Low HDL/High Triglycerides and Impact on Global Health Outcomes) trial. During a
Koon K Teo et al.
Stroke, 44(10), 2688-2693 (2013-07-25)
In Atherothrombosis Intervention in Metabolic Syndrome with low HDL/High Triglycerides: Impact on Global Health Outcomes (AIM-HIGH) trial, addition of extended-release niacin (ERN) to simvastatin in participants with established cardiovascular disease, low high-density lipoprotein cholesterol, and high triglycerides had no incremental
Paul M Lavigne et al.
Journal of the American College of Cardiology, 61(4), 440-446 (2012-12-26)
This study sought to assess the efficacy of niacin for reducing cardiovascular disease (CVD) events, as indicated by the aggregate body of clinical trial evidence including data from the recently published AIM-HIGH (Atherothrombosis Intervention in Metabolic Syndrome with Low HDL/High
Sorin Tunaru et al.
Nature medicine, 9(3), 352-355 (2003-02-04)
Nicotinic acid (niacin), a vitamin of the B complex, has been used for almost 50 years as a lipid-lowering drug. The pharmacological effect of nicotinic acid requires doses that are much higher than those provided by a normal diet. Its
Martina Lukasova et al.
Trends in pharmacological sciences, 32(12), 700-707 (2011-09-29)
Nicotinic acid (niacin) has been used for decades to prevent and treat atherosclerosis. The well-documented antiatherogenic activity is believed to result from its antidyslipidemic effects, which are accompanied by unwanted effects, especially a flush. There has been renewed interest in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service