Skip to Content
Merck
All Photos(1)

Documents

19-3310

Sigma-Aldrich

2-Butanone

SAJ first grade, ≥99.0%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

grade

SAJ first grade

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

960 °F

expl. lim.

10.1 %

availability

available only in Japan

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Application of adsorptive stripping voltammetry based on PbFE for the determination of trace aluminum released into the environment during the corrosion process: The study highlights the role of ammonia buffer solutions in environmental monitoring techniques, particularly in tracing metal ion activities affecting corrosion processes (Wasąg J, Grabarczyk M, 2021).

  • Long-Term Generation of Longitudinal Spin Order Controlled by Ammonia Ligation Enables Rapid SABRE Hyperpolarized 2D NMR: Demonstrates advanced NMR techniques facilitated by ammonia buffer solutions, enhancing molecular imaging capabilities essential in pharmaceutical research (Vaneeckhaute E, De Ridder S, Tyburn JM, Kempf JG, Taulelle F, Martens JA, Breynaert E, 2021).



Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

15.8 °F

Flash Point(C)

-9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S G Carmella et al.
Cancer research, 53(4), 721-724 (1993-02-15)
Metabolites of the tobacco-specific nitrosamine, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone, a potent pulmonary carcinogen, have been quantified in the urine of 11 smokers. They were not detected in nonsmokers' urine. The metabolites, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol and its glucuronide, were detected in quantities of 0.23-1.0 and 0.57-6.5
L B Goldsmith et al.
Contact dermatitis, 19(5), 348-350 (1988-11-01)
Irritant contact dermatitis along with an increased transepidermal water loss can result from exposing the skin to solvents. A study of the interaction of various solvents with human stratum corneum was made using thin-layer chromatography. Comparison of 10 solvents (trichloroethylene
Jeong-Hwan Yoon et al.
Nature communications, 6, 7600-7600 (2015-07-22)
Transforming growth factor-β (TGF-β) and interleukin-6 (IL-6) are the pivotal cytokines to induce IL-17-producing CD4(+) T helper cells (TH17); yet their signalling network remains largely unknown. Here we show that the highly homologous TGF-β receptor-regulated Smads (R-Smads): Smad2 and Smad3
Federico Mele et al.
Nature communications, 6, 6431-6431 (2015-03-17)
T helper (TH) cell polarization during priming is modulated by a number of signals, but whether polarization to a given phenotype also influences recall responses of memory TH cells is relatively unknown. Here we show that miR-181a is selectively induced
Sherille D Bradley et al.
Cancer immunology research, 3(6), 602-609 (2015-03-22)
Oncogene activation in tumor cells induces broad and complex cellular changes that contribute significantly to disease initiation and progression. In melanoma, oncogenic BRAF(V600E) has been shown to drive the transcription of a specific gene signature that can promote multiple mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service