Skip to Content
Merck
All Photos(1)

Key Documents

15-1560

Sigma-Aldrich

Isopentyl acetate

SAJ special grade, ≥98.0%

Synonym(s):

Acetic acid 3-methylbutyl ester, Isoamyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744750
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
SAJ special grade
bp:
142 °C/756 mmHg (lit.)
vapor pressure:
5 mmHg ( 25 °C)

grade

SAJ special grade

vapor density

4.5 (vs air)

vapor pressure

5 mmHg ( 25 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

680 °F

expl. lim.

7.5 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.4 (lit.)

bp

142 °C/756 mmHg (lit.)

mp

−78 °C (lit.)

density

0.876 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCOC(C)=O

InChI

1S/C7H14O2/c1-6(2)4-5-9-7(3)8/h6H,4-5H2,1-3H3

InChI key

MLFHJEHSLIIPHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Prud'homme et al.
Neuroscience, 162(4), 1287-1298 (2009-05-30)
Food odours are major determinants for food choice, and their detection depends on nutritional status. The effects of different odour stimuli on both behavioural responses (locomotor activity and sniffing) and Fos induction in olfactory bulbs (OB) were studied in satiated
Atsushi Ugajin et al.
PloS one, 7(3), e32902-e32902 (2012-03-21)
Anti-predator behaviors are essential to survival for most animals. The neural bases of such behaviors, however, remain largely unknown. Although honeybees commonly use their stingers to counterattack predators, the Japanese honeybee (Apis cerana japonica) uses a different strategy to fight
Dharmendra S Dheeman et al.
Bioresource technology, 102(3), 3373-3379 (2010-12-15)
An extracellular thermostable lipase from Amycolatopsis mediterranei DSM 43304 has been purified to homogeneity using ammonium sulphate precipitation followed by anion exchange chromatography and hydrophobic interaction chromatography. This protocol resulted in a 398-fold purification with 36% final recovery. The purified
Hiroshi Sahara et al.
Journal of bioscience and bioengineering, 108(5), 359-364 (2009-10-07)
By application of the high-efficiency loss of heterozygosity (HELOH) method for disrupting genes in diploid sake yeast (Kotaka et al., Appl. Microbiol. Biotechnol., 82, 387-395 (2009)), we constructed, from a heterozygous integrant, a homozygous diploid that overexpresses the alcohol acetyltransferase
A M Kleemann et al.
Physiology & behavior, 97(3-4), 401-405 (2009-03-24)
The human ability to localize odorants has been examined in a number of studies, but the findings are contradictory. In the present study we investigated the human sensitivity and ability to localize hydrogen sulphide (H(2)S), which in low concentrations stimulates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service