Skip to Content
Merck
All Photos(1)

Key Documents

12-1370

Sigma-Aldrich

Glyoxal solution

CP, 40 wt. % in H2O

Synonym(s):

Ethanedial, Oxalaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHCCHO
CAS Number:
Molecular Weight:
58.04
Beilstein:
1732463
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
CP

grade

CP

availability

available only in Japan

concentration

40 wt. % in H2O

dilution

(for analytical testing)

SMILES string

[H]C(=O)C([H])=O

InChI

1S/C2H2O2/c3-1-2-4/h1-2H

InChI key

LEQAOMBKQFMDFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.

Caution

May precipitate on storage; redissolve at 50-60 °C.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catherine Lambert et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 18(1), 59-69 (2012-10-30)
The molecular basis of Alzheimer's disease has not been clearly established, but disruption of brain metal ion homeostasis, particularly copper and zinc, might be closely involved in the pathogenesis of this disease and its characteristic β-amyloid neuropathological features. The use
Yu Wang et al.
Chemical Society reviews, 41(11), 4140-4149 (2012-04-18)
Methylglyoxal (MGO) and glyoxal (GO), known as reactive carbonyl species, can be generated endogenously and exogenously (human body and food system). They are attracting increased attention because of their relationship with diabetes and flavour generation. In this review, their characteristics
Fien Van Lancker et al.
Journal of agricultural and food chemistry, 60(18), 4697-4708 (2012-04-03)
Only a minor part of Maillard reaction studies in the literature focused on the reaction between carbohydrates and peptides. Therefore, in continuation of a previous study in which the influence of the peptide C-terminal amino acid was investigated, this study
Niklas Bosaeus et al.
Nucleic acids research, 42(12), 8083-8091 (2014-05-20)
Overstretching of DNA occurs at about 60-70 pN when a torsionally unconstrained double-stranded DNA molecule is stretched by its ends. During the transition, the contour length increases by up to 70% without complete strand dissociation. Three mechanisms are thought to
Sabrina Gensberger et al.
Analytical and bioanalytical chemistry, 403(10), 2923-2931 (2012-03-03)
High-fructose corn syrup (HFCS) is a widely used liquid sweetener produced from corn starch by hydrolysis and partial isomerization of glucose to fructose. During these processing steps, sugars can be considerably degraded, leading, for example, to the formation of reactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service