Skip to Content
Merck
All Photos(1)

Key Documents

07-3880

Sigma-Aldrich

Diisopropyl ether

JIS special grade, ≥99.0%

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

JIS special grade

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

827 °F

expl. lim.

1-21 %, 100 °F

availability

available only in Japan

refractive index

n20/D 1.367 (lit.)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.725 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Gonzalez-Olmos et al.
Chemosphere, 71(11), 2098-2105 (2008-02-27)
Methyl tert-butyl ether (MTBE) is the most widely used oxygenate in gasoline blending and has become one of the world's most widespread groundwater and surface water pollutants. Alternative oxygenates to MTBE, namely ethyl tert-butyl ether (ETBE), tert-amyl ether (TAME) and
B E Cham et al.
Journal of virological methods, 137(1), 160-163 (2006-07-01)
Many viruses including HIV, hepatitis C and hepatitis B, have an outer lipid envelope which maintains inserted viral peptides in the "correct" functional conformation and orientation. Disruption of the lipid envelope by most solvents destroys infectivity and often results in
P Adlercreutz
Biochimica et biophysica acta, 1163(2), 144-148 (1993-05-13)
Horse liver alcohol dehydrogenase and alpha-chymotrypsin were deposited on a porous support material, Celite. After equilibration at a well-defined water activity, the catalytic activity was measured with diisopropyl ether as reaction medium. The effects of the presence of polyols and
R van Gijn et al.
Journal of chromatography. B, Biomedical applications, 667(2), 269-276 (1995-05-19)
The staurosporine derivative, N-benzoylstaurosporine (CGP 41 251; I), is a protein kinase C inhibitor that has been selected for phase I clinical evaluation in cancer patients. We have developed a selective and sensitive assay of the drug and three potential
Fiorella Belpoggi et al.
Annals of the New York Academy of Sciences, 982, 70-86 (2003-02-04)
Tert-amyl-methyl ether (TAME) was administered by gavage in extra virgin olive oil solution at concentrations of 750, 250, or 0 mg/kg bw to groups of 100 male and 100 female Sprague-Dawley rats 8 weeks old at the start of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service