Skip to Content
Merck
All Photos(1)

Key Documents

05-6580

Sigma-Aldrich

Cresol red

JIS special grade

Synonym(s):

o-Cresolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O5S
CAS Number:
Molecular Weight:
382.43
Beilstein:
343399
EC Number:
MDL number:
UNSPSC Code:
12171502
PubChem Substance ID:

grade

JIS special grade

availability

available only in Japan

pH

7.2-8.8, yellow to red

SMILES string

Cc1cc(ccc1O)C2(OS(=O)(=O)c3ccccc23)c4ccc(O)c(C)c4

InChI

1S/C21H18O5S/c1-13-11-15(7-9-18(13)22)21(16-8-10-19(23)14(2)12-16)17-5-3-4-6-20(17)27(24,25)26-21/h3-12,22-23H,1-2H3

InChI key

OBRMNDMBJQTZHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Horie et al.
Analytical biochemistry, 137(1), 80-87 (1984-02-01)
A highly sensitive quantitative method was developed to detect protons released or taken up upon ligand binding. A small change in pH due to proton release or uptake was detected by measuring the difference in the absorbance of a pH
M Swati et al.
Analytica chimica acta, 676(1-2), 68-74 (2010-08-31)
An optical biosensing scheme based on urease encapsulated calcium alginate microspheres which are coated with polyelectrolyte nanofilms predominantly composed of cresol red (CR) dye is demonstrated in this paper. The dye molecules within the nanofilms are deposited via the layer-by-layer
Berthold Borucki et al.
Biochemistry, 41(31), 10026-10037 (2002-07-31)
We studied the kinetics of proton uptake and release by photoactive yellow protein (PYP) from Ectothiorhodospira halophila in wild type and the E46Q and E46A mutants by transient absorption spectroscopy with the pH-indicator dyes bromocresol purple or cresol red in
K Elamrani et al.
Biochimica et biophysica acta, 727(1), 22-30 (1983-01-05)
The kinetics of H+/OH- diffusion across dimyristoyl phosphatidic acid bilayer membranes was measured by following the absorbance of the pH-sensitive indicator Cresol red (o-cresolsulfonphthalein) entrapped in single lamellar vesicles after rapidly changing the external pH in a stopped-flow apparatus. The
K Schilling et al.
Clinical chemistry, 39(2), 251-256 (1993-02-01)
If buffer/indicator systems are used as optical thermometers in multicuvettes, the temperature resolution is limited by errors of optical measurements produced mainly by variations of pathlength and blank transmittance of the wells. Theoretical and practical approaches show that, in multicuvettes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service